missing translation for 'onlineSavingsMsg'
Learn More

1,6-Dibromohexane, 97+%

Product Code. 11495443
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495443 500 g 500g
11485443 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495443 Supplier Thermo Scientific Alfa Aesar Supplier No. A13417.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 629-03-8 | C6H12Br2 | 243.97 g/mol

1,6-Dibromohexane is used as a reagent in the synthesis of novel benzo[b]xanthone derivatives which have a potential antitumor activity. It is also used as a cross-linker for the cross-linking of glycuronans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,6-Dibromohexane is used as a reagent in the synthesis of novel benzo[b]xanthone derivatives which have a potential antitumor activity. It is also used as a cross-linker for the cross-linking of glycuronans.

Solubility
Miscible with ethanol, ether, benzene and chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-03-8
Molecular Formula C6H12Br2
Molecular Weight (g/mol) 243.97
MDL Number MFCD00000272
InChI Key SGRHVVLXEBNBDV-UHFFFAOYSA-N
Synonym hexamethylene dibromide, hexane, 1,6-dibromo, 1,6-dibromo-n-hexane, 1,6-dibromohexan, alpha,omega-dibromohexane, 1,6-dibromohexan german, hexamethylene bromide, labotest-bb ltbb001557, 1,6 dibromohexane
PubChem CID 12368
IUPAC Name 1,6-dibromohexane
SMILES C(CCCBr)CCBr

Specifications

Melting Point -2°C
Density 1.61
Boiling Point 242°C to 244°C
Flash Point >110°C (230°F)
Odor Odorless
Refractive Index 1.5075
Quantity 500 g
UN Number UN3082
Beilstein 1236322
Solubility Information Miscible with ethanol,ether,benzene and chloroform. Immiscible with water.
Formula Weight 243.98
Percent Purity ≥97%
Chemical Name or Material 1,6-Dibromohexane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.