missing translation for 'onlineSavingsMsg'
Learn More

1,6-Diiodohexane, 97+%, stab. with copper

Product Code. 11449150
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449150 100 g 100g
11439150 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449150 Supplier Thermo Scientific Alfa Aesar Supplier No. A15368.22
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 629-09-4 | C6H12I2 | 337.97 g/mol

1,6-Diiodohexane is used as a precursor in the preparation of cationic tissue plasminogen activator (t-PA)-containing polyurethane surfaces. It is also used as a solvent additive and in potential power conversion efficiency of polymer or fullerene bulk heterojunction solar cells. Further, it is employed in the preparation of 3-(iodohexyl)-1-(3-(triethoxysilyl)propylcarbamoyl)-1H-benzo[d]imidazol-3-ium iodide and 2,4-dihydroxybenzophenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,6-Diiodohexane is used as a precursor in the preparation of cationic tissue plasminogen activator (t-PA)-containing polyurethane surfaces. It is also used as a solvent additive and in potential power conversion efficiency of polymer or fullerene bulk heterojunction solar cells. Further, it is employed in the preparation of 3-(iodohexyl)-1-(3-(triethoxysilyl)propylcarbamoyl)-1H-benzo[d]imidazol-3-ium iodide and 2,4-dihydroxybenzophenone.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-09-4
Molecular Formula C6H12I2
Molecular Weight (g/mol) 337.97
MDL Number MFCD00001103
InChI Key QLIMAARBRDAYGQ-UHFFFAOYSA-N
Synonym hexamethylene diiodide, hexane, 1,6-diiodo, 1,6-diiodohexane, stabilized with copper, 1,6-di-iodohexane, 1,6-diiodo hexane, pubchem3846, hexamethyleneiodide, 1,6-bis iodanyl hexane, acmc-209tt7, ksc354g6d
PubChem CID 12373
IUPAC Name 1,6-diiodohexane
SMILES ICCCCCCI

Specifications

Density 2.052
Boiling Point 123°C to 128°C (4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5852
Quantity 100 g
Beilstein 1280128
Sensitivity Light sensitive
Solubility Information Immiscible with water.
Formula Weight 337.97
Percent Purity 98%
Chemical Name or Material 1,6-Diiodohexane, stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.