missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,6-Diiodoperfluorohexane, 97%

Product Code. 11399719
Change view
Click to view available options
Quantity:
2 g
5 g
25 g
Unit Size:
25g
2g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399719 5 g 5g
15403237 2 g 2g
11309729 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399719 Supplier Thermo Scientific Chemicals Supplier No. B21654.06
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 375-80-4 | C6F12I2 | 553.86 g/mol

It is employed in the synthesis and characterization of fluorinated telomers containing vinylidene fluoride and hexafluoropropene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis and characterization of fluorinated telomers containing vinylidene fluoride and hexafluoropropene.

Notes
Light sensitive. Protect from light. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,6-Diiodoperfluorohexane
CAS 375-80-4
Melting Point 25°C
Density 2.357
Boiling Point 115°C (100 mmHg)
Molecular Formula C6F12I2
Refractive Index 1.398
MDL Number MFCD00042264
Quantity 5 g
Beilstein 1822186
Sensitivity Light sensitive
Synonym 1,6-diiodoperfluorohexane, 1,6-diiodododecafluorohexane, dodecafluoro-1,6-diiodohexane, 1,6-diiododecafluorohexane, perfluoro-1,6-diiodohexane, hexane, 1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodo, acmc-209iu0, 1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodo-hexane, hexane,1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodo
InChI Key JOQDDLBOAIKFQX-UHFFFAOYSA-N
SMILES FC(F)(I)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I
IUPAC Name 1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodohexane
Molecular Weight (g/mol) 553.86
PubChem CID 78994
Formula Weight 553.86
Percent Purity 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.