missing translation for 'onlineSavingsMsg'
Learn More

1,6-Diiodoperfluorohexane, TRC

Product Code. 30383473
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383473 250 mg
30378404 5 g
30378539 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383473 Supplier Toronto Research Chemicals Supplier No. TRCD492508250MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375-80-4
Molecular Formula C6F12I2
Molecular Weight (g/mol) 553.85
Synonym 1,1,2,2,3,3,4,4,5,5,6,6-Dodecafluoro-1,6-diiodohexane, 1,6-Diiodododecafluorohexane, 1,6-Diiodoperfluorohexane, 1,6-Iododecafluorohexane, Dodecafluoro-1,6-diiodohexane, alpha,ω-Diiodoperfluorohexane
SMILES FC(F)(I)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C6F12I2/c7-1(8,3(11,12)5(15,16)19)2(9,10)4(13,14)6(17,18)20
Formula Weight 553.7898
Chemical Name or Material 1,6-Diiodoperfluorohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.