missing translation for 'onlineSavingsMsg'
Learn More

1,6-Hexanediol diacrylate, 99% (reactive esters), stab. with 90ppm hydroquinone

Product Code. 11339268
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339268 250 g 250g
11349268 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339268 Supplier Thermo Scientific Alfa Aesar Supplier No. 043203.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 13048-33-4 | C12H18O4 | 226.27 g/mol

1,6-Hexanediol diacrylate is used as a functional monomer for polymers. It acts as a cross linking agent between the molecular chains of polymers. Further, it is used in adhesives, sealants, alkyd coatings, elastomers, photopolymers, and inks for improved adhesion, hardness, abrasion and heat resistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13048-33-4
Molecular Formula C12H18O4
Molecular Weight (g/mol) 226.27
MDL Number MFCD00008631
InChI Key FIHBHSQYSYVZQE-UHFFFAOYSA-N
Synonym 1,6-hexanediol diacrylate, hexamethylene diacrylate, kayarad hdda, hexane-1,6-diyl diacrylate, 1,6-hexamethylene diacrylate, hexamethylene glycol diacrylate, sartomer 238, hdoda, viscoat 230, hexamethylene acrylate
PubChem CID 25644
IUPAC Name 6-prop-2-enoyloxyhexyl prop-2-enoate
SMILES C=CC(=O)OCCCCCCOC(=O)C=C

Specifications

Density 1.010 g/mL
Flash Point >110°C (>230°F)
Quantity 250 g
Sensitivity Hygroscopic
Solubility Information Slightly miscible with water.
Formula Weight 226.28
Percent Purity 99%
Physical Form Liquid
Assay (reactive esters)
Chemical Name or Material 1,6-Hexanediol diacrylate, Stabilized with 90ppm hydroquinone
compliance-icons
Product Identifier
  • 1,6-Hexanediol diacrylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1,6-Hexanediol diacrylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.