missing translation for 'onlineSavingsMsg'
Learn More

1,6-Hexanediol Diacrylate (stabilized with 100 ppm MEHQ) (>85%), TRC

Product Code. 30136372
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136372 500 mg 500mg
30012613 5 g 5g
17954815 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136372 Supplier Toronto Research Chemicals Supplier No. TRCH294253500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13048-33-4
Molecular Formula C12H18O4
Molecular Weight (g/mol) 226.27
Synonym 1,2-Bisacryloyloxyhexane, 1,6-HDDA, 1,6-Hexamethylene Diacrylate, 1,6-Hexamethylenediol Diacrylate, 1,6-Hexandiol Diacrylate, 1,6-Hexanediol Diacrylate, 1,6-Hexanediyl Diacrylate, 1,6-Hexenediol Diacrylate, 1,6-Hexyleneglycol Diacrylate, 1,6HX-A, 1.6HX, A-HD-N, AHD, Actilane 425, AgiSyn 2816, C 6DA;C 716, Cognis 4017, Doublemer HDDA, EM 2021, EM 221, EM 221TF, FA 126AS;Fancryl FA 126AS, Genomer 1223, HDDA, HDODA, Hexamethylene Acrylate;Hexamethylene Diacrylate, Hexamethylenediol Diacrylate, Hexamethyleneglycol Diacrylate, Hexaneglycol Diacrylate, Hexylene-1,6-diacrylate, KS-HDDA, Kayarad HDDA, Kayarad KS-HDDA, LM 254, Laromer HDDA, Light Acrylate 1,6HX-A;Light Acrylate SR 238F, Light Ester 1.6HX-A, M 200, M 200 (acrylate), Miramer M 200, NK Ester A-HD, NK Ester A-HD-N, Newfrontier HDDA, Photomer 4017;Photomer 4017F, SM 626, SR 238, SR 2389, SR 238B, SR 238F, SR 238NS;SR 238TFN, Sartomer 238, Sartomer SR 238, Setalux UV 2243, V 230, Viscoat 230;
IUPAC Name 6-prop-2-enoyloxyhexyl prop-2-enoate
SMILES C=CC(=O)OCCCCCCOC(=O)C=C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C12H18O4/c1-3-11(13)15-9-7-5-6-8-10-16-12(14)4-2/h3-4H,1-2,5-10H2
Formula Weight 226.1205
Chemical Name or Material 1,6-Hexanediol Diacrylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.