missing translation for 'onlineSavingsMsg'
Learn More

1,6-Hexanediol, TRC

Product Code. 30403170
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30403170 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30403170 Supplier Toronto Research Chemicals Supplier No. TRCH290098500MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 629-11-8
Molecular Formula C6 H14 O2
Quantity 500 mg
Synonym 1,6-Dihydroxyhexane, 1,6-Hexylene glycol, Hexamethylene glycol, Hexamethylenediol, NSC 508, α,ω-Hexanediol, ω-Hexanediol
InChI Formula InChI=1S/C6H14O2/c7-5-3-1-2-4-6-8/h7-8H,1-6H2
SMILES OCCCCCCO
IUPAC Name hexane-1,6-diol
Molecular Weight (g/mol) 118.17
Formula Weight 118.0994
Chemical Name or Material 1,6-Hexanediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.