missing translation for 'onlineSavingsMsg'
Learn More

[1-(6-Methylpyrazin-2-yl)piperid-3-yl]methanol, 95%, Thermo Scientific™

Product Code. 10749415
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10749415 250 mg Amber Glass Bottle 250mg
10163424 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10749415 Supplier Thermo Scientific Maybridge Supplier No. CC59009CB
 Restricted Item
Estimated Shipment: 09-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 937795-91-0
Molecular Formula C11H17N3O
Molecular Weight (g/mol) 207.277
MDL Number MFCD09879916
InChI Key OWOMYFSSKZGDBC-UHFFFAOYSA-N
Synonym 1-6-methylpyrazin-2-yl piperid-3-yl methanol, 3-hydroxymethyl-1-6-methylpyrazin-2-yl piperidine, 1-6-methylpyrazin-2-yl piperidin-3-yl methanol, 1-6-methylpyrazin-2-yl-3-piperidyl methan-1-ol
PubChem CID 24229598
IUPAC Name [1-(6-methylpyrazin-2-yl)piperidin-3-yl]methanol
SMILES CC1=CN=CC(=N1)N2CCCC(C2)CO

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 207.28
Percent Purity 95%
Chemical Name or Material [1-(6-Methylpyrazin-2-yl)piperid-3-yl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.