missing translation for 'onlineSavingsMsg'
Learn More

1-[6-(Trifluoromethyl)pyridin-2-yl]piperazine, 97%, Thermo Scientific™

Product Code. 10574984
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10574984 1 g Amber Glass Bottle 1g
10152754 250 mg Amber Glass Bottle 250mg
10555860 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10574984 Supplier Thermo Scientific Maybridge Supplier No. SEW03804DA
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 127561-18-6
Molecular Formula C10H12F3N3
Molecular Weight (g/mol) 231.222
InChI Key IAOMYUZAXMFANI-UHFFFAOYSA-N
Synonym 1-6-trifluoromethyl pyridin-2-yl piperazine, 2-piperazino-6-trifluoromethyl pyridine, 1-6-trifluoromethyl-2-pyridinyl piperazine, piperazine, 1-6-trifluoromethyl-2-pyridinyl, 6-trifluoromethyl-2-pyridyl piperazine, acmc-209bcl, 4-6-trifluoromethyl-2-pyridyl piperazine, 1-6-trifluoromethyl-2-pyridyl piperazine
PubChem CID 11424740
IUPAC Name 1-[6-(trifluoromethyl)pyridin-2-yl]piperazine
SMILES C1CN(CCN1)C2=CC=CC(=N2)C(F)(F)F

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 231.22
Percent Purity 97%
Chemical Name or Material 1-[6-(trifluoromethyl)pyridin-2-yl]piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.