missing translation for 'onlineSavingsMsg'
Learn More

1,7-Dioxaspiro[5.5]undecane, 98%

Product Code. 11460237
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460237 100 g 100g
11440237 5 g 5g
11450237 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460237 Supplier Thermo Scientific Alfa Aesar Supplier No. B21664.22
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 180-84-7 | C9H16O2 | 156.225 g/mol

Suitable for use in organic farming.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Suitable for use in organic farming.

Solubility
Soluble in water 1.0g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 180-84-7
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD00011578
InChI Key GBBVHDGKDQAEOT-UHFFFAOYSA-N
Synonym 1,7-dioxaspiro 5.5 undecane, olean, unii-khi3qeq4zj, 1,7-dioxaspiro 5,5 undecane, khi3qeq4zj, r-1,7-dioxaspiro 5.5 undecane, s-1,7-dioxaspiro 5.5 undecane, olive-fly ketal, eco-trap
PubChem CID 67437
IUPAC Name 1,7-dioxaspiro[5.5]undecane
SMILES C1CCOC2(C1)CCCCO2

Spécification

Density 1.02
Boiling Point 193°C
Flash Point 63°C (145°F)
Refractive Index 1.464
Quantity 100 g
Beilstein 105997
Solubility Information Soluble in water 1.0g/L.
Formula Weight 156.23
Percent Purity 98%
Chemical Name or Material 1,7-Dioxaspiro[5.5]undecane

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.