missing translation for 'onlineSavingsMsg'
Learn More

1,7-Dioxaspiro[5.5]undecane, TRC

Product Code. 30372825
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372825 5 g -
30373505 1 g 1g
30382743 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30372825 Supplier Toronto Research Chemicals Supplier No. TRCD4863255G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 180-84-7
Molecular Formula C9 H16 O2
Molecular Weight (g/mol) 156.22
Synonym 1,7-Dioxaspiro[5.5]undecane, Olean
IUPAC Name 1,7-dioxaspiro[5.5]undecane
SMILES C1CCC2(CCCCO2)OC1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H16O2/c1-3-7-10-9(5-1)6-2-4-8-11-9/h1-8H2
Formula Weight 156.115
Chemical Name or Material 1,7-Dioxaspiro[5.5]undecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.