missing translation for 'onlineSavingsMsg'
Learn More

1,7-Heptanediol, 95%

Product Code. 10494535 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10494535 1 g Glass bottle 1g
10346750 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10494535 Supplier Thermo Scientific Acros Supplier No. 120350010
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 629-30-1 | C7H16O2 | 132.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-30-1
Molecular Formula C7H16O2
Molecular Weight (g/mol) 132.2
MDL Number MFCD00002987
InChI Key SXCBDZAEHILGLM-UHFFFAOYSA-N
Synonym 1,7-heptanediol, 1,7-dihydroxyheptane, heptamethylene glycol, alpha,omega-heptanediol, .alpha.,.omega.-heptanediol, a,?-heptanediol, 1,7-heptandiol, .omega.-heptanediol, pubchem11489, acmc-1bsor
PubChem CID 12381
IUPAC Name heptane-1,7-diol
SMILES C(CCCO)CCCO

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 17°C to 19°C
Color Colorless to Yellow
Density 0.9510g/mL
Boiling Point 259°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula HO(CH2)7OH
Refractive Index 1.4535 to 1.4565
Quantity 1 g
Beilstein 01, 489
Specific Gravity 0.951
Formula Weight 132.2
Percent Purity 95%
Physical Form Liquid After Melting
Chemical Name or Material 1, 7-Heptanediol, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.