missing translation for 'onlineSavingsMsg'
Learn More

1,8-Diaminooctane, 98%

Product Code. 11436736
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436736 100 g 100g
11426736 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11436736 Supplier Thermo Scientific Alfa Aesar Supplier No. B23885.22
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 373-44-4 | C8H22Cl2N2 | 217.18 g/mol

1,8-Diaminooctane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Diaminooctane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water 575g/L.

Notes
Air sensitive & hygroscopic. Incompatible with oxidizing agents, carbon dioxide, air, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 373-44-4
Molecular Formula C8H22Cl2N2
Molecular Weight (g/mol) 217.18
MDL Number MFCD00008248
InChI Key ZFLWZOGXFQNIMT-UHFFFAOYSA-N
Synonym 1,8-diaminooctane, 1,8-octanediamine, octamethylenediamine, 1,8-octylenediamine, 1,8-octamethylenediamine, diaminooctane, octane 1,8-diamine, unii-53a6694pie, alpha,omega-diaminooctane, chembl29392
PubChem CID 24250
ChEBI CHEBI:73112
IUPAC Name octane-1,8-diamine
SMILES [Cl-].[Cl-].[NH3+]CCCCCCCC[NH3+]

Specifications

Melting Point 50°C to 54°C
pH 11.7
Boiling Point 225°C to 226°C
Flash Point 106°C (223°F)
Odor Amine-like
Quantity 100 g
UN Number UN3259
Beilstein 1735426
Sensitivity Air sensitive; Hygroscopic
Solubility Information Soluble in water 575g/L.
Formula Weight 144.26
Percent Purity 98%
Chemical Name or Material 1,8-Diaminooctane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,8-Diaminooctane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Octamethylenediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.