missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,8-Dihydroxyanthraquinone, 95%
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 191°C to 193°C (sublimation) |
| Quantity | 100 g |
| Beilstein | 2054727 |
| Merck Index | 14,2815 |
| Formula Weight | 240.21 |
| Percent Purity | 95% |
| Chemical Name or Material | 1,8-Dihydroxyanthraquinone |
Safety and Handling
Product Identifier
- 1,8-Dihydroxyanthraquinone
- Warning
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : 1,8-Dihydroxyanthraquinone
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction