missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dihydroxyanthraquinone, 95%

Product Code. 11494044
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11494044

Brand: Thermo Scientific Alfa Aesar A12001.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 117-10-2 | C14H8O4 | 240.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-10-2
Molecular Formula C14H8O4
Molecular Weight (g/mol) 240.214
MDL Number MFCD00001211
InChI Key QBPFLULOKWLNNW-UHFFFAOYSA-N
Synonym danthron, 1,8-dihydroxyanthraquinone, chrysazin, dantron, antrapurol, laxanthreen, diaquone, istizin, laxanorm, altan
PubChem CID 2950
ChEBI CHEBI:3682
IUPAC Name 1,8-dihydroxyanthracene-9,10-dione
SMILES C1=CC2=C(C(=C1)O)C(=O)C3=C(C2=O)C=CC=C3O

Specifications

Melting Point 191°C to 193°C (sublimation)
Quantity 500 g
Beilstein 2054727
Merck Index 14,2815
Formula Weight 240.21
Percent Purity 95%
Chemical Name or Material 1,8-Dihydroxyanthraquinone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.