missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dioxacyclotetradecane-2,9-dione, TRC

Product Code. 30381504
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30381504 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381504 Supplier Toronto Research Chemicals Supplier No. TRCD42681550MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,8-Dioxacyclotetradecane-2,9-dione
CAS 1020-83-3
Molecular Formula C12 H20 O4
Quantity 50 mg
Synonym 2-Oxepanone, dimer, Caprolactone cyclic dimer, Dicaprolactone, ε-Caprolactone cyclic dimer
InChI Formula InChI=1S/C12H20O4/c13-11-7-3-1-5-9-15-12(14)8-4-2-6-10-16-11/h1-10H2
SMILES O=C1CCCCCOC(=O)CCCCCO1
IUPAC Name 1,8-dioxacyclotetradecane-2,9-dione
Molecular Weight (g/mol) 228.285
Formula Weight 228.136
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.