missing translation for 'onlineSavingsMsg'
Learn More

1,8-Octanediol, 98+%

Product Code. 11498366
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498366 100 g 100g
11488366 25 g 25g
11408376 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498366 Supplier Thermo Scientific Alfa Aesar Supplier No. A15402.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 629-41-4 | C8H18O2 | 146.23 g/mol

1,8-Octanediol is used in cosmetics, perfumes, ink, essence and in UV coating. It acts as a precursor in the synthesis of polymer and in the manufacture of pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Octanediol is used in cosmetics, perfumes, ink, essence and in UV coating. It acts as a precursor in the synthesis of polymer and in the manufacture of pharmaceuticals.

Solubility
Soluble in water and methanol.

Notes
Incompatible with strong oxidizing agents, reducing agents, acid chlorides acid, anhydrides and chloroformates.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-41-4
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00002989
InChI Key OEIJHBUUFURJLI-UHFFFAOYSA-N
PubChem CID 69420
ChEBI CHEBI:44630
IUPAC Name octane-1,8-diol
SMILES C(CCCCO)CCCO

Specifications

Melting Point 58°C to 61°C
Boiling Point 171°C to 173°C (20 mmHg)
Flash Point 148°C (298°F)
Assay Percent Range ≥98%
Linear Formula HO(CH2)8OH
Quantity 100 g
Beilstein 1633499
Synonym 1,8-octanediol, octamethylene glycol, 1,8-dihydroxyoctane, octan-1,8-diol, 1,8-octandiol, 1, 8-octandiol, octane-1,8-diol;, acmc-209nan, ksc358i9p
Solubility Information Soluble in water and methanol.
Formula Weight 146.23
Percent Purity ≥98%
Chemical Name or Material 1,8-Octanediol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.