missing translation for 'onlineSavingsMsg'
Learn More

1,8-Octanediol, 98%

Product Code. 15299626 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15299626 100 g 100g
15219626 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15299626 Supplier Thermo Scientific Acros Supplier No. 129381000
 Restricted Item
Estimated Shipment: 22-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 629-41-4 | C8H18O2 | 146.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-41-4
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
InChI Key OEIJHBUUFURJLI-UHFFFAOYSA-N
PubChem CID 69420
ChEBI CHEBI:44630
IUPAC Name octane-1,8-diol
SMILES C(CCCCO)CCCO

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 60.0°C to 62.0°C
Color White to Yellow
Boiling Point 278.8°C
Flash Point 128°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula HO(CH2)8OH
Quantity 100 g
Beilstein 01, 490
Synonym 1,8-octanediol, octamethylene glycol, 1,8-dihydroxyoctane, octan-1,8-diol, 1,8-octandiol, 1, 8-octandiol, octane-1,8-diol;, acmc-209nan, ksc358i9p
Solubility Information Solubility in water: slightly soluble in water.
Viscosity 20.6 mPa.s (70°C)
Formula Weight 146.23
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 1, 8-Octanediol, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 1,8-Octanediol, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.