missing translation for 'onlineSavingsMsg'
Learn More

1,9-Decadiyne, 97%

Product Code. 11484924
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484924 5 g 5g
11494924 25 g 25g
11404934 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484924 Supplier Thermo Scientific Alfa Aesar Supplier No. A13834.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1720-38-3 | C10H14 | 134.22 g/mol

One-step synthesis of carbametacyclophane 3 from 1,9-decadiyne and diethyl acetylenedicarboxylate is described. The use of 1,9-decadiyne as a nucleophile led only to the monoalkynylation product, a selectivity that would be difficult to achieve with a catalytic in situ metalation process. 1,9-decadiyne reacts with iodobenzene (excess) to afford the bis-Sonogashira coupling product in excellent yield. 1,9-Decadiyne was dihydrated and gave 2,9-decanedione in 92% yield.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
One-step synthesis of carbametacyclophane 3 from 1,9-decadiyne and diethyl acetylenedicarboxylate is described. The use of 1,9-decadiyne as a nucleophile led only to the monoalkynylation product, a selectivity that would be difficult to achieve with a catalytic in situ metalation process. 1,9-decadiyne reacts with iodobenzene (excess) to afford the bis-Sonogashira coupling product in excellent yield. 1,9-Decadiyne was dihydrated and gave 2,9-decanedione in 92% yield.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1720-38-3
Molecular Formula C10H14
Molecular Weight (g/mol) 134.22
MDL Number MFCD00014926
InChI Key ILVDYAGPHFWNQI-UHFFFAOYSA-N
Synonym 1,9-decadiyne, acmc-1bppd, 1,9-decadiyne 6ci, 7ci, 8ci, 9ci
PubChem CID 74400
ChEBI CHEBI:37823
IUPAC Name deca-1,9-diyne
SMILES C#CCCCCCCC#C

Specifications

Density 0.822
Boiling Point 80°C to 81°C (10 mmHg)
Flash Point 60°C (140°F)
Refractive Index 1.451
Quantity 5 g
UN Number UN3295
Beilstein 1734594
Solubility Information Insoluble in water.
Formula Weight 134.22
Percent Purity 97%
Chemical Name or Material 1,9-Decadiyne
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,9-Decadiyne
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1,9-Decadiyne

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.