missing translation for 'onlineSavingsMsg'
Learn More

1,9-Dimethyl-methylene Blue Zinc Chloride Double Salt (Technical Grade), TRC

Product Code. 30380412
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380412 100 mg
30373412 500 mg
30387006 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380412 Supplier Toronto Research Chemicals Supplier No. TRCD469355100MG
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 931418-92-7
Molecular Formula 2 C18 H22 N3 S . 2Cl . Zn Cl2
Molecular Weight (g/mol) 832.12
Synonym 1,9-Dimethyl-Methylene Blue Zinc Chloride Double Salt
IUPAC Name bis(3,7-bis(dimethylamino)-1,9-dimethylphenothiazin-5-ium) zinc(II) chloride
SMILES CC1=C2C([S+]=C(C3=N2)C=C(C=C3C)N(C)C)=CC(N(C)C)=C1.CC4=C5C([S+]=C(C6=N5)C=C(C=C6C)N(C)C)=CC(N(C)C)=C4.Cl[Zn]Cl.[Cl-].[Cl-]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/2C18H22N3S.4ClH.Zn/c2*1-11-7-13(20(3)4)9-15-17(11)19-18-12(2)8-14(21(5)6)10-16(18)22-15;;;;;/h2*7-10H,1-6H3;4*1H;/q2*+1;;;;;+2/p-4
Formula Weight 832.09
Chemical Name or Material 1,9-Dimethyl-methylene Blue Zinc Chloride Double Salt (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.