missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-4-piperidinecarboxylic acid, 99%, Thermo Scientific™

Product Code. 10602135 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10602135 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10602135 Supplier Thermo Scientific Acros Supplier No. 194630050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25503-90-6
Molecular Formula C8H12NO3
Molecular Weight (g/mol) 170.19
MDL Number MFCD00023706
InChI Key WFCLWJHOKCQYOQ-UHFFFAOYSA-M
Synonym 1-acetyl-4-piperidinecarboxylic acid, 1-acetylisonipecotic acid, 1-acetyl-4-piperidine carboxylic acid, 4-piperidinecarboxylic acid, 1-acetyl, n-acetyl-4-piperidinecarboxylic acid, 1-acetyl-piperidine-4-carboxylic acid, maybridge1_003128, 1-acetylpiperidine-4-carboxylic acid,, acetylisonipecotic acid, acmc-209gkl
PubChem CID 117255
SMILES CC(=O)N1CCC(CC1)C([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 180.0°C to 182.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: soluble in methanol, ethanol, benzene and toluene
IUPAC Name 1-acetylpiperidine-4-carboxylic acid
Formula Weight 171.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1-Acetyl-4-piperidinecarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 1-Acetyl-4-piperidinecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.