missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylguanidine, 97%

Product Code. 10245114 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10245114 5 g Glass bottle 5g
10081364 1 g Glass bottle 1g
10774375 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10245114 Supplier Thermo Scientific Acros Supplier No. 397440050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5699-40-1 | C3H7N3O | 101.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5699-40-1
Molecular Formula C3H7N3O
Molecular Weight (g/mol) 101.11
MDL Number MFCD00075615
InChI Key NGGXACLSAZXJGM-UHFFFAOYSA-N
Synonym n-acetylguanidine, 1-acetylguanidine, n-carbamimidoylacetamide, n-aminoiminomethyl acetamide, monoacetyl guanidine, acetylguanidine, n-acetyl-guanidine, acetyl guanidine, n-diaminomethylidene acetamide, 1-acetylguanidne
PubChem CID 38263
IUPAC Name N-(diaminomethylidene)acetamide
SMILES CC(=O)N=C(N)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 181.0°C to 183.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 101.11
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1-Acetylguanidine
Show More Show Less
compliance-icons
Product Identifier
  • 1-Acetylguanidine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.