missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylpiperidine-4-carboxylic acid, 98+%

Product Code. 11437477
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11437477 5 g 5g
11447477 25 g 25g
11457477 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11437477 Supplier Thermo Scientific Alfa Aesar Supplier No. A15606.06
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 25503-90-6 | C8H12NO3 | 170.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25503-90-6
Molecular Formula C8H12NO3
Molecular Weight (g/mol) 170.19
MDL Number MFCD00023706
InChI Key WFCLWJHOKCQYOQ-UHFFFAOYSA-M
Synonym 1-acetyl-4-piperidinecarboxylic acid, 1-acetylisonipecotic acid, 1-acetyl-4-piperidine carboxylic acid, 4-piperidinecarboxylic acid, 1-acetyl, n-acetyl-4-piperidinecarboxylic acid, 1-acetyl-piperidine-4-carboxylic acid, maybridge1_003128, 1-acetylpiperidine-4-carboxylic acid,, acetylisonipecotic acid, acmc-209gkl
PubChem CID 117255
IUPAC Name 1-acetylpiperidine-4-carboxylic acid
SMILES CC(=O)N1CCC(CC1)C([O-])=O

Specifications

Melting Point 180°C to 184°C
Quantity 5 g
Beilstein 389577
Formula Weight 171.19
Percent Purity ≥98%
Chemical Name or Material 1-Acetylpiperidine-4-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.