missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantaneacetonitrile, 97%

Product Code. 11466496
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11466496

Brand: Thermo Scientific Alfa Aesar B24770.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16269-13-9 | C12H17N | 175.28 g/mol

1-Adamantaneacetonitrile is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantaneacetonitrile is used as pharmaceutical intermediate.

Solubility
Sparingly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16269-13-9
Molecular Formula C12H17N
Molecular Weight (g/mol) 175.28
MDL Number MFCD00194034
InChI Key DXQVPXCZIRQITG-UHFFFAOYSA-N
Synonym 2-adamantan-1-yl acetonitrile, 1-adamantaneacetonitrile, 1-adamantylacetonitrile, 2-1-adamantyl acetonitrile, tricyclo 3.3.1.13,7 decane-1-acetonitrile, 2-adamantanylethanenitrile, pubchem21521, 1-adamantylmethyl cyanide, adamantane-1-acetonitrile
PubChem CID 140052
SMILES N#CCC12CC3CC(CC(C3)C1)C2

Specifications

Quantity 1 g
UN Number UN3439
Solubility Information Sparingly soluble in water.
IUPAC Name 2-(1-adamantyl)acetonitrile
Formula Weight 175.28
Percent Purity 97%
Chemical Name or Material 1-Adamantaneacetonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.