missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantyl Bromide, TRC

Product Code. 17938444
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17938444 1 g 1g
30134356 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17938444 Supplier Toronto Research Chemicals Supplier No. TRCA2085001G
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 768-90-1
Molecular Formula C10 H15 Br
Molecular Weight (g/mol) 215.13
Synonym 1-Bromo-tricyclo[3.3.1.13,7]decane, 1-Bromoadamantane, 1-Bromoadamantane, NSC 527914
IUPAC Name 1-bromoadamantane
SMILES C12(Br)CC3CC(CC(C3)C1)C2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H15Br/c11-10-4-7-1-8(5-10)3-9(2-7)6-10/h7-9H,1-6H2/t7-,8+,9-,10-
Formula Weight 214.0357
Chemical Name or Material 1-Adamantyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.