missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantyl isocyanate, 98%

Product Code. 10607114 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10607114 1 g Glass bottle 1g
10452841 5 g Glass bottle 5g
10531364 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10607114 Supplier Thermo Scientific Acros Supplier No. 314360010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4411-25-0 | C11H15NO | 177.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4411-25-0
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.25
MDL Number MFCD00134405
InChI Key VBHCPGFCIQDXGZ-UHFFFAOYSA-N
Synonym 1-adamantyl isocyanate, 1-isocyanato-adamantane, 1-isocyantoadamantane, 1-adamantylisocyanate, adamantanisocyanate, adamantyl isocyanate, pubchem8848, 1-adamantyl-isocyanate, 1-adamantanyl isocyanate, adamantane, 1-isocyanato
PubChem CID 2059995
IUPAC Name 1-isocyanatoadamantane
SMILES O=C=NC12CC3CC(CC(C3)C1)C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 144.0°C to 146.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 177.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-Adamantyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.