missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-1-cyclohexanecarboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 10491051 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
Product Code. Quantity Packaging unitSize
10491051 25 g Glass bottle 25g
10114460 100 g Glass bottle 100g
2 options
This item is not returnable. View return policy

Product Code. 10491051

Brand: Thermo Scientific Acros 203950250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2756-85-6
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.19
MDL Number MFCD00001487
InChI Key WOXWUZCRWJWTRT-UHFFFAOYSA-N
Synonym 1-aminocyclohexanecarboxylic acid, homocycloleucine, 1-amino-1-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 1-amino, alpha-aminocyclohexanecarboxylic acid, alpha-aminocyclohexanecarboxylic aicd, unii-1se3hd391h, 1-amino-1-cyclohexane carboxylic acid, .alpha.-aminocyclohexanecarboxylic acid, .alpha.-aminocyclohexanecarboxylic aicd
PubChem CID 1366
ChEBI CHEBI:86534
IUPAC Name 1-aminocyclohexane-1-carboxylic acid
SMILES NC1(CCCCC1)C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >300°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 14, 299
Formula Weight 143.19
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 1-Amino-1-cyclohexanecarboxylic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Amino-1-cyclohexanecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.