missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-11-azido-3,6,9-trioxaundecane, Thermo Scientific Chemicals

Product Code. 15476309
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15476309 100 mg 100mg
15466309 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. Consulta la politica di reso
Codice prodotto. 15476309 Fornitore Thermo Scientific Alfa Aesar N. del fornitore J64308.MC
 Restricted Item
In Stock
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 134179-38-7 | Molecular Formula: C8H18N4O3 | Molecular Weight: 218.257 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 134179-38-7
Molecular Formula C8H18N4O3
Molecular Weight (g/mol) 218.257
MDL Number MFCD00269874
InChI Key FPVCVHVTMPCZTH-UHFFFAOYSA-N
Synonym 1-amino-11-azido-3,6,9-trioxaundecane, azido-peg3-amine, 11-azido-3,6,9-trioxaundecan-1-amine, 2-2-2-2-azidoethoxy ethoxy ethoxy ethanamine, 1-2-2-aminoethoxy ethoxy-2-2-azidoethoxy ethane, 11-azido-3 6 9-trioxaundecan-1-amine, ethanamine, 2-2-2-2-azidoethoxy ethoxy ethoxy, 2-2-2-2-azidoethoxy ethoxy ethoxy ethylamine
PubChem CID 2735280
IUPAC Name 2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethanamine
SMILES C(COCCOCCOCCN=[N+]=[N-])N

Specifica

Color Brown to Yellow
Linear Formula N3CH2CH2(OCH2CH2)3NH2
Quantity 100 mg
UN Number UN2735
Formula Weight 218.26
Physical Form Viscous Liquid
Chemical Name or Material 1-Amino-11-azido-3,6,9-trioxaundecane

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.