missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-3-(9H-carbazol-4-yloxy)-2-propanol, TRC

Product Code. 30326274
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326274 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326274 Supplier Toronto Research Chemicals Supplier No. TRCA611075500MG
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Amino-3-(9H-carbazol-4-yloxy)-2-propanol
CAS 72955-96-5
Molecular Formula C15 H16 N2 O2
Quantity 500 mg
Synonym 2-Propanol, 1-amino-3-(9H-carbazol-4-yloxy)-, 1-Amino-3-(9H-carbazol-4-yloxy)-2-propanol, 1-Amino-3-(9H-carbazol-4-yloxy)propan-2-ol
SMILES NCC(O)COc1cccc2[nH]c3ccccc3c12
Molecular Weight (g/mol) 256.3
Formula Weight 256.1212
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.