missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-4-methylpiperazine, 98%

Product Code. 11369645
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11369645 25 g 25g
11379645 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11369645 Supplier Thermo Scientific Alfa Aesar Supplier No. L19108.14
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 6928-85-4 | C5H13N3 | 115.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6928-85-4
Molecular Formula C5H13N3
Molecular Weight (g/mol) 115.18
MDL Number MFCD00006154
InChI Key RJWLLQWLBMJCFD-UHFFFAOYSA-N
Synonym 1-amino-4-methylpiperazine, 1-piperazinamine, 4-methyl, 4-methyl-1-piperazinamine, 4-methyl-piperazin-1-ylamine, 1-methyl-4-aminopiperazine, 4-amino-1-methylpiperazine, 4-methylpiperazinylamine, 1-methyl-4-aminopierazine, pubchem8577, n-amino-4-methylpiperazine
PubChem CID 81349
IUPAC Name 4-methylpiperazin-1-amine
SMILES CN1CCN(CC1)N

Specifications

Density 0.96
Boiling Point 172°C to 175°C
Flash Point 70°C (158°F)
Refractive Index 1.484
Quantity 25 g
UN Number UN2735
Beilstein 103334
Sensitivity Air and light sensitive
Formula Weight 115.18
Percent Purity 98%
Chemical Name or Material 1-Amino-4-methylpiperazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.