missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Amino-7-naphthalenesulfonic Acid, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | 1-Amino-7-naphthalenesulfonic Acid |
| CAS | 119-28-8 |
| Molecular Formula | C10H9NO3S |
| Quantity | 25 g |
| Synonym | 8-Amino-2-naphthalenesulfonic Acid, 1-Naphthylamine-7-sulfonic Acid, 1-Amino-7-sulfonaphthalene, 1,7-Cleve’s Acid |
| InChI Formula | InChI=1S/C10H9NO3S/c11-10-3-1-2-7-4-5-8(6-9(7)10)15(12,13)14/h1-6H,11H2,(H,12,13,14) |
| SMILES | Nc1cccc2ccc(cc12)S(=O)(=O)O |
| IUPAC Name | 8-aminonaphthalene-2-sulfonic acid |
| Molecular Weight (g/mol) | 223.25 |
| Formula Weight | 223.0303 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction