Learn More
1-Aminobenzotriazole, 97%
Description
1-Aminobenzotriazole is used as a Suicide inhibitor of cytochrome P450 and chloroperoxidase, an inhibitor of oxidative drug metabolism in biochemical pharmacology and toxicology studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Aminobenzotriazole is used as a Suicide inhibitor of cytochrome P450 and chloroperoxidase, an inhibitor of oxidative drug metabolism in biochemical pharmacology and toxicology studies.
Solubility
Soluble in chloroform, DMSO, ethanol or water
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Specifications
| Melting Point | 81°C to 85°C |
| Color | Brown to White |
| Quantity | 1 g |
| Beilstein | 607843 |
| Solubility Information | Soluble in chloroform, DMSO, ethanol or water |
| Formula Weight | 134.14 |
| Percent Purity | 98% |
| Physical Form | Powder |
| Chemical Name or Material | 1-Aminobenzotriazole |
Safety and Handling
- 1-Aminobenzotriazole
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : 1-Aminobenzotriazole
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.