missing translation for 'onlineSavingsMsg'
Learn More

1-Aminobenzotriazole, 97%

Product Code. p-7042743
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15475799 250 mg 250mg
15455799 1 g 1g
15465799 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15475799 Supplier Thermo Scientific Alfa Aesar Supplier No. J63610.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Suicide inhibitor of cytochrome P450 and chloroperoxidase | CAS: 1614-12-6 | C6H6N4 | 134.142 g/mol

1-Aminobenzotriazole is used as a Suicide inhibitor of cytochrome P450 and chloroperoxidase, an inhibitor of oxidative drug metabolism in biochemical pharmacology and toxicology studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Aminobenzotriazole is used as a Suicide inhibitor of cytochrome P450 and chloroperoxidase, an inhibitor of oxidative drug metabolism in biochemical pharmacology and toxicology studies.

Solubility
Soluble in chloroform, DMSO, ethanol or water

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1614-12-6
Molecular Formula C6H6N4
Molecular Weight (g/mol) 134.142
MDL Number MFCD00132902
InChI Key JCXKHYLLVKZPKE-UHFFFAOYSA-N
Synonym 1-aminobenzotriazole, 1h-benzotriazol-1-amine, 1h-benzo d 1,2,3 triazol-1-amine, 1-benzotriazolamine, 1-abtz, 1h-1,2,3-benzotriazol-1-amine, 1h-1,2,3-benzotriazol-1-ylamine, 1,2,3-benzotriazol-1-amine, benzotriazolylamine, 1-benzotriazolylamine
PubChem CID 1367
IUPAC Name benzotriazol-1-amine
SMILES C1=CC=C2C(=C1)N=NN2N

Specifications

Melting Point 81°C to 85°C
Color Brown to White
Quantity 250 mg
Beilstein 607843
Solubility Information Soluble in chloroform, DMSO, ethanol or water
Formula Weight 134.14
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-Aminobenzotriazole
compliance-icons
Product Identifier
  • 1-Aminobenzotriazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 1-Aminobenzotriazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.