missing translation for 'onlineSavingsMsg'
Learn More

1-Aminocyclopropane-1-carboxylic Acid, TRC

Product Code. 30134933
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134933 1 g 1g
30133707 5 g 5g
30007881 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134933 Supplier Toronto Research Chemicals Supplier No. TRCA6035501G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22059-21-8
Molecular Formula C4 H7 N O2
Molecular Weight (g/mol) 101.1
Synonym Cyclopropanecarboxylic acid, 1-amino- (6CI, 7CI, 8CI, 9CI, ACI), 1-Aminocyclopropanecarboxylic acid (ACI), 1-Amino-1-carboxycyclopropane, 1-Aminocyclopropane-1-carboxylate, 1-Aminocyclopropane-1-carboxylic acid, ACC, ACPC, NSC 98430, α-Aminocyclopropanecarboxylic acid, Cyclopropanecarboxylic acid, 1-amino- (6CI, 7CI, 8CI, 9CI, ACI), 1-Aminocyclopropanecarboxylic acid (ACI), 1-Amino-1-carboxycyclopropane, 1-Aminocyclopropane-1-carboxylate, 1-Aminocyclopropane-1-carboxylic acid, ACC, ACPC, NSC 98430, α-Aminocyclopropanecarboxylic acid
IUPAC Name 1-aminocyclopropane-1-carboxylic acid
SMILES NC1(CC1)C(=O)O

Specifications

InChI Formula InChI=1S/C4H7NO2/c5-4(1-2-4)3(6)7/h1-2,5H2,(H,6,7)
Quantity 1 g
Formula Weight 101.0477
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material 1-Aminocyclopropane-1-carboxylic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.