missing translation for 'onlineSavingsMsg'
Learn More

1-Aminohydantoin hydrochloride, 90%

Product Code. 11956251 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11956251 5 g Glass bottle 5g
11966251 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11956251 Supplier Thermo Scientific Acros Supplier No. 442050050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2827-56-7 | C3H5N3O2·ClH | 151.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2827-56-7
Molecular Formula C3H5N3O2·ClH
Molecular Weight (g/mol) 151.55
InChI Key WEOHANUVLKERQI-UHFFFAOYSA-N
Synonym 1-aminohydantoin hydrochloride, 1-aminoimidazolidine-2,4-dione hydrochloride, 1-aminohydantoin hcl, 1-amino hydantoin hydrochloride, 2,4-imidazolidinedione, 1-amino-, monohydrochloride, acmc-209h1m, 1-aminoimidazolidine-2,4-dione hcl, 1-aminoimidazoline-2,4-dione hydrochloride
PubChem CID 12472963
IUPAC Name 1-aminoimidazolidine-2,4-dione;hydrochloride
SMILES C1C(=O)NC(=O)N1N.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 201.0°C to 205.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (Argentometry)
Packaging Glass bottle
Quantity 5 g
Formula Weight 151.55
Percent Purity 90%
Physical Form Crystalline Powder
Chemical Name or Material 1-Aminohydantoin hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.