missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Aminopyrene-d9, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 1-Aminopyrene-d9 |
| CAS | 128008-86-6 |
| Molecular Formula | C16 D9 H2 N |
| Quantity | 0.01 g |
| Synonym | 1-Pyren-2,3,4,5,6,7,8,9,10-d9-amine (9 CI), 1-Aminopyrene-d9, 1-Aminopyrene-D9 |
| InChI Formula | InChI=1 S/C16H11N/c17-14-9-7-12-5-4-10-2-1-3-11-6-8-13(14)16(12)15(10)11/h1-9 H,17H2/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D |
| SMILES | [2 H]c1c([2 H])c2c([2 H])c([2 H])c3c([2 H])c([2 H])c(N)c4c([2 H])c([2 H])c(c1[2 H])c2c34 |
| IUPAC Name | 2,3,4,5,6,7,8,9,10-nonadeuteriopyren-1-amine |
| Molecular Weight (g/mol) | 226.321 |
| Formula Weight | 226.146 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction