missing translation for 'onlineSavingsMsg'
Learn More

1-Benzo[b]thiophen-3-yl-2-bromoethan-1-one, 97%, Thermo Scientific™

Product Code. 11905558
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11905558 5 g Amber Glass Bottle 5g
10120732 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11905558 Supplier Thermo Scientific Maybridge Supplier No. CC12305DE
 Restricted Item
Estimated Shipment: 15-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 26167-45-3
Molecular Formula C10H7BrOS
Molecular Weight (g/mol) 255.129
InChI Key CKHWNGWAHFLCTJ-UHFFFAOYSA-N
Synonym 1-benzo b thiophen-3-yl-2-bromoethanone, 1-benzo b thiophen-3-yl-2-bromoethan-1-one, 3-bromoacetyl benzothiophene, 1-1-benzothiophen-3-yl-2-bromoethan-1-one, 3-bromoacetyl benzo b thiophene, 1-1-benzothiophen-3-yl-2-bromoethanone, 1-1-benzothiophen-3-yl-2-bromo-1-ethanone, 3-2-bromoacetyl benzo b thiophen, 1-benzothiophen-3-yl-2-bromo-ethanone, ethanone,1-benzo b thien-3-yl-2-bromo
PubChem CID 2776341
IUPAC Name 1-(1-benzothiophen-3-yl)-2-bromoethanone
SMILES C1=CC=C2C(=C1)C(=CS2)C(=O)CBr

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 255.13
Percent Purity 97%
Chemical Name or Material 1-Benzo[b]thiophen-3-yl-2-bromoethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.