missing translation for 'onlineSavingsMsg'
Learn More

1-Benzofuran-5-ylmethanol, Thermo Scientific™

Product Code. p-4509958
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10056012 250 mg Amber Glass Bottle 250mg
10670135 1 g Amber Glass Bottle 1g
10285164 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10056012 Supplier Thermo Scientific Maybridge Supplier No. CC07109CB
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 31823-05-9
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
MDL Number MFCD04115373
InChI Key XSLXZYDPOMAXTM-UHFFFAOYSA-N
Synonym benzofuran-5-ylmethanol, 5-benzofuranmethanol, 5-hydroxymethylbenzofuran, pubchem7026, 5-hydroxymethyl-benzofuran, 5-hydroxymethylbenzo b furan, benzofuran-5-methanol, 1-benzofuran-5-yl methanol, 1-benzo b furan-5-ylmethanol, benzo b furan-5-ylmethan-1-ol
PubChem CID 2795179
IUPAC Name 1-benzofuran-5-ylmethanol
SMILES OCC1=CC=C2OC=CC2=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 148.16
Percent Purity 97%
Chemical Name or Material 1-Benzofuran-5-ylmethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.