missing translation for 'onlineSavingsMsg'
Learn More

1-Benzothiophen-3-ylmethylamine hydrochloride, 95%, Thermo Scientific™

Product Code. 11340567
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11340567 5 g 5g
10723043 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11340567 Supplier Thermo Scientific Maybridge Supplier No. CC12313DE
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 55810-74-7
Molecular Formula C9H10ClNS
Molecular Weight (g/mol) 199.696
MDL Number MFCD01566652
InChI Key MIAPJPCDRSPNPW-UHFFFAOYSA-N
Synonym benzo b thiophen-3-ylmethanamine hydrochloride, 1-benzothiophen-3-ylmethanamine hydrochloride, 1-benzothiophen-3-ylmethylamine hydrochloride, benzo b thiophen-3-ylmethylamine hydrochloride, 1-1-benzothiophen-3-yl methanamine hydrochloride, benzo b thiophen-3-ylmethylamine, chloride, 1-benzothiophen-3-yl methanamine hydrochloride, 3-aminomethyl benzo b thiophene hydrochloride, benzo b thiophen-3-yl-methylamine hydrochloride salt, 1-1-benzothiophen-3-yl methanamine-hydrogen chloride 1/1
PubChem CID 2776346
IUPAC Name 1-benzothiophen-3-ylmethanamine;hydrochloride
SMILES C1=CC=C2C(=C1)C(=CS2)CN.Cl

Specifications

CAS Min % 80.0
CAS Max % 100.0
Assay Percent Range 95%
Quantity 5 g
Formula Weight 199.7
Percent Purity 95%
Chemical Name or Material 1-Benzothiophen-3-ylmethylamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.