missing translation for 'onlineSavingsMsg'
Learn More

1-Benzothiophen-5-yl isocyanate, 97%, Thermo Scientific™

Product Code. 10788245
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10788245 5 g 5g
10771114 250 mg 250mg
10196252 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10788245 Supplier Thermo Scientific Maybridge Supplier No. CC24806DE
 Restricted Item
Estimated Shipment: 15-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 239097-78-0
Molecular Formula C9H5NOS
Molecular Weight (g/mol) 175.205
InChI Key GWLZJAWTWVSHCU-UHFFFAOYSA-N
Synonym 1-benzothiophen-5-yl isocyanate, 1-benzothiophen-5-ylisocyanate, benzo b thiophen-5-isocyanate, 5-isocyanato-benzo b thiophene, benzo b thiophene,5-isocyanato
PubChem CID 7060535
IUPAC Name 5-isocyanato-1-benzothiophene
SMILES C1=CC2=C(C=CS2)C=C1N=C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 175.21
Percent Purity 97%
Chemical Name or Material 1-Benzothiophen-5-yl isocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.