missing translation for 'onlineSavingsMsg'
Learn More

1-Benzothiophene-2-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10772463
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10772463 1 g Amber Glass Bottle 1g
10304712 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10772463 Supplier Thermo Scientific Maybridge Supplier No. CC12204DA
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 3541-37-5
Molecular Formula C9H6OS
Molecular Weight (g/mol) 162.21
MDL Number MFCD01075041
InChI Key NXSVNPSWARVMAY-UHFFFAOYSA-N
Synonym benzo b thiophene-2-carboxaldehyde, benzo b thiophene-2-carbaldehyde, thianaphthene-2-carboxaldehyde, benzothiophene-2-carboxaldehyde, acmc-209igo, 2-formylbenzo b thiophene, 2-benzothiophenecarbaldehyde, 2-formyl-benzo b thiophene, benzothiophene-2-carbaldehyde, 2-benzothiophenecarboxaldehyde
PubChem CID 736500
IUPAC Name 1-benzothiophene-2-carbaldehyde
SMILES O=CC1=CC2=CC=CC=C2S1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula Yes
Quantity 1 g
Formula Weight 162.21
Percent Purity 97%
Chemical Name or Material 1-Benzothiophene-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.