missing translation for 'onlineSavingsMsg'
Learn More

1-Benzothiophene-2-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10314662
Change view
Click to view available options
Amber Glass Bottle; 250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10314662 Amber Glass Bottle; 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10314662 Supplier Thermo Scientific Maybridge Supplier No. CC12203CB
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 90001-64-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H5ClO2S2
MDL Number MFCD03659701
Quantity 250 mg
Synonym benzo b thiophene-2-sulfonyl chloride, 1-benzothiophene-2-sulfonylchloride, benzo b thiophene-2-sulfonylchloride, benzothiophene-2-sulfonyl chloride, pubchem10221, 2-benzothiophenesulfonyl chloride, benzo b thiophen-2-ylchlorosulfone, 1-benzothiophene-2-sulphonyl chloride, 2-chlorosulphonyl-1-benzothiophene, 1-benzo b thiophene-2-sulfonyl chloride
InChI Key FKIIVBOPAHICHQ-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C=C(S2)S(=O)(=O)Cl
IUPAC Name 1-benzothiophene-2-sulfonyl chloride
Molecular Weight (g/mol) 232.696
PubChem CID 2776333
Formula Weight 232.71
Percent Purity 97%
Chemical Name or Material 1-Benzothiophene-2-sulfonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.