missing translation for 'onlineSavingsMsg'
Learn More

1-Benzothiophene-3-carboximidamidine hydrochloride hydrate, 97%, Thermo Scientific™

Product Code. 10191012
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10191012 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10191012 Supplier Thermo Scientific Maybridge Supplier No. CC12331EA
 Restricted Item
Estimated Shipment: 05-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 465515-36-0
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H11ClN2OS
MDL Number MFCD03659730
Quantity 10 g
Synonym 1-benzothiophene-3-carboximidamide hydrochloride, 1-benzothiophene-3-carboximidamidine hydrochloride, benzo b thiophene-3-carboxamidine, chloride
InChI Key DCDNRDYUSJNDGA-UHFFFAOYSA-N
SMILES O.Cl.NC(=N)C1=CSC2=CC=CC=C12
IUPAC Name 1-benzothiophene-3-carboximidamide;hydrate;hydrochloride
Molecular Weight (g/mol) 230.71
PubChem CID 2776350
Formula Weight 212.7
Percent Purity 97%
Chemical Name or Material 1-Benzothiophene-3-carboximidamidine hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.