missing translation for 'onlineSavingsMsg'
Learn More

1-Benzoylcyclohexanol, TRC

Product Code. 30331667
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30331667 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331667 Supplier Toronto Research Chemicals Supplier No. TRCB2084185G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Benzoyl-1-hydroxycyclohexane
CAS 947-19-3
Molecular Formula C13 H16 O2
Quantity 5 g
Synonym Ketone, 1-hydroxycyclohexyl phenyl (6CI,7CI,8CI), (1-Hydroxycyclohexyl)phenylmethanone, 1-Benzoyl-1-hydroxycyclohexane, 1-Benzoylcyclohexanol, 1-Hydroxy-1-cyclohexyl phenyl ketone, 1-Hydroxycyclohexyl phenyl ketone, 184D, Additol CPK, BASF 184, Boxing 184, CPK, Chemcure 481, Chivacure 184, Ciba 184, Daido UV-Cure 174, Darocur 184C, Doublecure 184, Esacure KS 300, Genocure CPK, HCPK, Huatai 184, I 184, I 189, IC 184, IHT-PI 184, IRG 184, Initiator 1104, Irgacure 1104, Irgacure 181, Irgacure 183, Irgacure 184, Irgacure 1841, Irgacure 184D, Irgacure 189, Irgacure G 17-1184, Irgacure I 184, JR 184, JRCure 1104, JRcure 184, KS 300, Luna 200, Lunacure 200, Micure CP 4, NSC 401908, Omnirad 184, Omnirad 481, PI 184, Photocure 184, Runtecure 1104, SB-PI 714, SR 1122, SarCure SR 1122, Speedcure 184, Suncure 84, UV 184, X-Cure 184, photoinitiator 184, α-Hydroxy-α-cyclohexyl phenyl ketone, α-Hydroxycyclohexyl phenyl ketone
InChI Formula InChI=1S/C13H16O2/c14-12(11-7-3-1-4-8-11)13(15)9-5-2-6-10-13/h1,3-4,7-8,15H,2,5-6,9-10H2
SMILES OC1(CCCCC1)C(=O)c2ccccc2
IUPAC Name (1-hydroxycyclohexyl)-phenylmethanone
Molecular Weight (g/mol) 204.26
Formula Weight 204.115
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.