missing translation for 'onlineSavingsMsg'
Learn More

1-Benzoylpiperazine, 97%

Product Code. 15452288
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15452288 1 g 1g
15462288 5 g 5g
15472288 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15452288 Supplier Thermo Scientific Alfa Aesar Supplier No. H55896.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 13754-38-6 | C11H14N2O | 190.246 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13754-38-6
Molecular Formula C11H14N2O
Molecular Weight (g/mol) 190.246
MDL Number MFCD00810192
InChI Key VUNXBQRNMNVUMV-UHFFFAOYSA-N
Synonym 1-benzoylpiperazine, n-benzoylpiperazine, phenyl-piperazin-1-yl-methanone, 1-benzoyl piperazine, 1-benzoyl-piperazine, phenyl piperazin-1-yl methanone, piperazine, 1-benzoyl, phenyl piperazinyl ketone, phenyl piperazin-4-ium-1-yl methanone, 1-phenylcarbonyl piperazine
PubChem CID 762654
IUPAC Name phenyl(piperazin-1-yl)methanone
SMILES C1CN(CCN1)C(=O)C2=CC=CC=C2

Specifications

Melting Point 61°C to 64°C
Boiling Point 145°C (0.05 mmHg)
Quantity 1 g
Formula Weight 190.24
Percent Purity 97%
Chemical Name or Material 1-Benzoylpiperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.