missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-1H-indol-5-ylamine, 97%, Thermo Scientific™

Product Code. 10563232
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10563232 10 g Amber Glass Bottle 10g
10416862 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10563232 Supplier Thermo Scientific Maybridge Supplier No. MO00835EA
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 26807-73-8
Molecular Formula C15H14N2
Molecular Weight (g/mol) 222.291
MDL Number MFCD03070173
InChI Key UYDNPZLYDODKKA-UHFFFAOYSA-N
Synonym 1-benzyl-1h-indol-5-ylamine, 1-benzyl-1h-indol-5-amine, 5-amino-1-benzylindole, n-benzyl-5-amino indole, 1-benzylindole-5-ylamine, 5-amino-1benzyl-1h-indole, 5-amino-1-benzyl-1h-indole, 1h-indol-5-amine,1-phenylmethyl
PubChem CID 2794624
IUPAC Name 1-benzylindol-5-amine
SMILES C1=CC=C(C=C1)CN2C=CC3=C2C=CC(=C3)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 222.29
Percent Purity 97%
Chemical Name or Material 1-Benzyl-1H-indol-5-ylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.