missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-2-(methylsulfanyl)-1H-imidazole-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10471483
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10471483

Brand: Thermo Scientific Maybridge CC28904DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 479400-30-1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C12H12N2OS
MDL Number MFCD04974046
Quantity 1 g
Synonym 1-benzyl-2-methylsulfanyl-1h-imidazole-5-carbaldehyde, 3-benzyl-2-methylsulfanyl imidazole-4-carbaldehyde, 2-methylthio-1-benzylimidazole-5-carbaldehyde, 1-benzyl-2-methylthio-1h-imidazole-5-carbaldehyde, 2-methylthio-3-benzyl-3h-imidazole-4-carbaldehyde
InChI Key FELACOPTUQTZRP-UHFFFAOYSA-N
SMILES CSC1=NC=C(N1CC2=CC=CC=C2)C=O
IUPAC Name 3-benzyl-2-methylsulfanylimidazole-4-carbaldehyde
Molecular Weight (g/mol) 232.301
PubChem CID 343414
Formula Weight 232.31
Percent Purity 97%
Chemical Name or Material 1-Benzyl-2-(methylsulfanyl)-1H-imidazole-5-carbaldehyde
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.