missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-2-(methylsulfanyl)-1H-imidazole-5-carbonylchloride hydrochloride, ≥97%, Thermo Scientific™

Product Code. 10117462
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10117462

Brand: Thermo Scientific Maybridge CC28902DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 568577-86-6
Molecular Formula C12H12Cl2N2OS
Molecular Weight (g/mol) 303.201
MDL Number MFCD04974045
InChI Key AUZZRVMDIAHPNZ-UHFFFAOYSA-N
Synonym 1-benzyl-2-methylsulfanyl-1h-imidazole-5-carbonyl chloride hydrochloride, 3-benzyl-2-methylsulfanyl imidazole-4-carbonyl chloride hydrochloride, 3-benzyl-2-methylsulfanylimidazole-4-carbonyl chloride hydrochloride, 1-benzyl-2-methylthio-1h-imidazole-5-carbonyl chloride hydrochloride
PubChem CID 2795486
IUPAC Name 3-benzyl-2-methylsulfanylimidazole-4-carbonyl chloride;hydrochloride
SMILES CSC1=NC=C(N1CC2=CC=CC=C2)C(=O)Cl.Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 303.21
Percent Purity 97%
Chemical Name or Material 1-Benzyl-2-(methylsulfanyl)-1H-imidazole-5-carbonyl chloride hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.