missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-2-(methylsulfanyl)-1H-imidazole-5-carbonylchloride hydrochloride, ≥97%, Thermo Scientific™

Product Code. 10178394
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10178394 5 g 5g
10117462 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10178394 Supplier Thermo Scientific Maybridge Supplier No. CC28902DE
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 568577-86-6
Molecular Formula C12H12Cl2N2OS
Molecular Weight (g/mol) 303.201
MDL Number MFCD04974045
InChI Key AUZZRVMDIAHPNZ-UHFFFAOYSA-N
Synonym 1-benzyl-2-methylsulfanyl-1h-imidazole-5-carbonyl chloride hydrochloride, 3-benzyl-2-methylsulfanyl imidazole-4-carbonyl chloride hydrochloride, 3-benzyl-2-methylsulfanylimidazole-4-carbonyl chloride hydrochloride, 1-benzyl-2-methylthio-1h-imidazole-5-carbonyl chloride hydrochloride
PubChem CID 2795486
IUPAC Name 3-benzyl-2-methylsulfanylimidazole-4-carbonyl chloride;hydrochloride
SMILES CSC1=NC=C(N1CC2=CC=CC=C2)C(=O)Cl.Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 303.21
Percent Purity 97%
Chemical Name or Material 1-Benzyl-2-(methylsulfanyl)-1H-imidazole-5-carbonyl chloride hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.