missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-2-pyrrolidinone, 97%, Thermo Scientific Chemicals

Product Code. 11495547
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495547 100 g 100g
11485547 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495547 Supplier Thermo Scientific Alfa Aesar Supplier No. B23538.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

1-Benzyl-2-pyrrolidinone undergoes reduction in THF with lithium N,N-dialkylaminoborohydrides to yield N-benzylpyrrolidine. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Benzyl-2-pyrrolidinone undergoes reduction in THF with lithium N,N-dialkylaminoborohydrides to yield N-benzylpyrrolidine. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5291-77-0
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.231
MDL Number MFCD00003195
InChI Key LVUQCTGSDJLWCE-UHFFFAOYSA-N
Synonym 1-benzyl-2-pyrrolidinone, n-benzyl-2-pyrrolidinone, n-benzyl-2-pyrrolidone, 1-benzyl-2-pyrrolidone, n-benzylpyrrolidin-2-one, 2-pyrrolidinone, 1-phenylmethyl, n-benzylpyrrolidone, 1-benzylpyrrolidone, n-benzylpyrrolidinone, acmc-1aste
PubChem CID 79176
IUPAC Name 1-benzylpyrrolidin-2-one
SMILES C1CC(=O)N(C1)CC2=CC=CC=C2

Specifications

Density 1.095
Boiling Point 76°C to 78°C (0.1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.552
Quantity 100 g
Beilstein 132168
Solubility Information Slightly soluble in water.
Formula Weight 175.23
Percent Purity 97%
Chemical Name or Material 1-Benzyl-2-pyrrolidinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.